Gene Transcripts

Gene Symbol
ZNRF1
Entrez Gene ID
84937
Gene Name
zinc and ring finger 1, E3 ubiquitin protein ligase
Gene Aliases
NIN283
Chromosome Location
Chr. 16: 74999017 - 75110994 on Build GRCh38
UniGene ID
Hs.427284
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032268.4 NP_115644.1 2, 3 1177
XM_017023793.1 XP_016879282.1 2, 3 1151
GenBank mRNA AF378524.1 - 2, 3 849
AK294434.1 - 2, 3 849
AL136903.1 - 2, 3 842
BC007235.1 - 2, 3 849 2823677

View Full Product Details