Gene Transcripts

Gene Symbol
C1orf94
Entrez Gene ID
84970
Gene Name
chromosome 1 open reading frame 94
Gene Aliases
Chromosome Location
Chr. 1: 34166883 - 34219131 on Build GRCh38
UniGene ID
Hs.194610
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134734.1 NP_001128206.1 6, 7 2559
NM_032884.4 NP_116273.2 6, 7 1690
GenBank mRNA AA725646.1 - 1, 2 443
AK123355.1 - 6, 7 1690
BC007637.1 - 6, 7 1458 3529463
BC064845.1 - 6, 7 1841 30390219

View Full Product Details