Gene Transcripts

Gene Symbol
TMEM41A
Entrez Gene ID
90407
Gene Name
transmembrane protein 41A
Gene Aliases
2900010K02Rik
Chromosome Location
Chr. 3: 185482671 - 185499057 on Build GRCh38
UniGene ID
Hs.677570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080652.3 NP_542383.1 5 758
XM_017007437.1 XP_016862926.1 4 596
GenBank mRNA AK290878.1 - 5 736
AK313954.1 - 5 694
AL832782.1 - 2 217
AY358547.1 - 5 742
BC008043.2 - 5 715 3838280
BC019884.1 - 5 705 4991416
BX647128.1 - 3 408

View Full Product Details