Gene Transcripts

Gene Symbol
DEPDC7
Entrez Gene ID
91614
Gene Name
DEP domain containing 7
Gene Aliases
TR2, dJ85M6.4
Chromosome Location
Chr. 11: 33015864 - 33033582 on Build GRCh38
UniGene ID
Hs.280990
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077242.1 NP_001070710.1 5 981
NM_139160.2 NP_631899.2 5 1220
GenBank mRNA AF447588.1 - 5 1220
AJ245600.1 - 5 936
BC030970.1 - 5 981 4133262

View Full Product Details