Gene Transcripts

Gene Symbol
SNX18
Entrez Gene ID
112574
Gene Name
sorting nexin 18
Gene Aliases
SH3PX2, SH3PXD3B, SNAG1
Chromosome Location
Chr. 5: 54517759 - 54619249 on Build GRCh38
UniGene ID
Hs.432755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102575.1 NP_001096045.1 1 1060
NM_001145427.1 NP_001138899.1 1 1064
NM_052870.2 NP_443102.2 1 1060
XM_017008997.1 XP_016864486.1 1 1064
GenBank mRNA AF395536.1 - 1 870
AK304195.1 - 1 1064
BC060791.1 - 1 1060 30341956
BC067860.1 - 1 1060 30343220
BC117218.1 - 1 958 40125769
BC117220.1 - 1 958 40125771

View Full Product Details