Gene Transcripts

Gene Symbol
IZUMO4
Entrez Gene ID
113177
Gene Name
IZUMO family member 4
Gene Aliases
C19orf36, IMAGE:4215339
Chromosome Location
Chr. 19: 2096848 - 2099590 on Build GRCh38
UniGene ID
Hs.424049
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031735.2 NP_001026905.2 1 175
NM_001039846.1 NP_001034935.1 1 175
XM_005259480.3 XP_005259537.2 1 196
GenBank mRNA AK294083.1 - 1 46
AK301957.1 - 1 129
AY358444.1 - 1 175
AY753639.1 - 1 134
BC010443.1 - 1 114
BC014609.2 - 1 129 4825119
BC040964.1 - 1 116
BI827974.1 - 1 32
BX378854.2 - 1 175
DQ196309.1 - 1 162
DQ196310.1 - 1 162
DQ424900.1 - 1 98

View Full Product Details