Gene Transcripts

Gene Symbol
GLCCI1
Entrez Gene ID
113263
Gene Name
glucocorticoid induced 1
Gene Aliases
FAM117C, GCTR, GIG18, TSSN1
Chromosome Location
Chr. 7: 7968743 - 8089080 on Build GRCh38
UniGene ID
Hs.131673
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138426.3 NP_612435.1 5, 6 1564
GenBank mRNA AL833694.1 - 1, 2 59
BC050291.1 - 5, 6 1515 5272003

View Full Product Details