Gene Transcripts

Gene Symbol
DDIT4L
Entrez Gene ID
115265
Gene Name
DNA damage inducible transcript 4 like
Gene Aliases
REDD2, Rtp801L
Chromosome Location
Chr. 4: 100185870 - 100190498 on Build GRCh38
UniGene ID
Hs.480378
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145244.3 NP_660287.1 3 795
GenBank mRNA AK094236.1 - 3 765
AK312926.1 - 3 765
AL832277.1 - 3 747
BC013592.2 - 3 753 3877854

View Full Product Details