Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052943.3 | NP_443175.2 | 1 | 368 | |
GenBank mRNA | AK122816.1 | - | 1 | 498 | |
BC012790.2 | - | 1 | 362 | 3957388 | |
BC014160.2 | - | 1 | 362 | 4542896 | |
CN311438.1 | - | 1 | 368 |