Gene Transcripts

Gene Symbol
TLCD1
Entrez Gene ID
116238
Gene Name
TLC domain containing 1
Gene Aliases
Chromosome Location
Chr. 17: 28724348 - 28727930 on Build GRCh38
UniGene ID
Hs.499952
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160407.1 NP_001153879.1 2 215
NM_138463.3 NP_612472.1 2 348
XM_006721671.3 XP_006721734.1 2 151
XM_011524278.2 XP_011522580.1 2 1791
GenBank mRNA BC014072.2 - 2 344
CA453962.1 - 2 354

View Full Product Details