Gene Transcripts

Gene Symbol
PIK3AP1
Entrez Gene ID
118788
Gene Name
phosphoinositide-3-kinase adaptor protein 1
Gene Aliases
BCAP
Chromosome Location
Chr. 10: 96593312 - 96720522 on Build GRCh38
UniGene ID
Hs.310456
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152309.2 NP_689522.2 14, 15 2297
XM_005269498.1 XP_005269555.1 13, 14 1820
XM_005269499.1 XP_005269556.1 13, 14 1844
XM_011539248.1 XP_011537550.1 14, 15 2289
GenBank mRNA AK122584.1 - 3, 4 3126
AK125635.1 - 14, 15 2289
BC029917.1 - 7, 8 1148 5168493
BX648550.1 - 14, 15 2193

View Full Product Details