Gene Transcripts

Gene Symbol
DNAJC24
Entrez Gene ID
120526
Gene Name
DnaJ heat shock protein family (Hsp40) member C24
Gene Aliases
DPH4, JJJ3, ZCSL3
Chromosome Location
Chr. 11: 31369829 - 31432835 on Build GRCh38
UniGene ID
Hs.187269
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181706.4 NP_859057.4 2 184
XM_017017216.1 XP_016872705.1 2 169
GenBank mRNA AK123087.1 - 2 175
AK289665.1 - 2 172
AL833128.1 - 2 174
BC036571.1 - 2 159 4793909
BC044931.2 - 2 139
BC063804.1 - 2 127 6474523
CD685768.1 - 2 202

View Full Product Details