Gene Transcripts

Gene Symbol
MISP
Entrez Gene ID
126353
Gene Name
mitotic spindle positioning
Gene Aliases
C19orf21, MISP1
Chromosome Location
Chr. 19: 748413 - 764319 on Build GRCh38
UniGene ID
Hs.439180
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173481.3 NP_775752.1 2 473
XM_011527685.2 XP_011525987.1 2 518
XM_011527686.2 XP_011525988.1 2 611
GenBank mRNA BC042125.2 - 2 440
BC052236.1 - 2 460 5431360
BX648389.1 - 2 454
HY163020.1 - 2 475

View Full Product Details