Gene Transcripts

Gene Symbol
C19orf47
Entrez Gene ID
126526
Gene Name
chromosome 19 open reading frame 47
Gene Aliases
Chromosome Location
Chr. 19: 40292743 - 40348527 on Build GRCh38
UniGene ID
Hs.631557
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256440.1 NP_001243369.1 7 648
NM_001256441.1 NP_001243370.1 7 643
NR_046202.1 7 696
XM_005258520.2 XP_005258577.1 7 637
XM_011526460.2 XP_011524762.1 7 636
XM_017026291.1 XP_016881780.1 7 648
XM_017026292.1 XP_016881781.1 6 557
XM_017026293.1 XP_016881782.1 7 648
XM_017026294.1 XP_016881783.1 4 366
XM_017026295.1 XP_016881784.1 8 746
GenBank mRNA AK094207.1 - 7 636
AL834131.1 - 7 673
BC023623.2 - 7 674 4662735
BC027935.1 - 7 664 5202287
BI916290.1 - 7 643
BM459040.1 - 7 593
CR936760.1 - 7 649

View Full Product Details