Gene Transcripts

Gene Symbol
UBXN10
Entrez Gene ID
127733
Gene Name
UBX domain protein 10
Gene Aliases
UBXD3
Chromosome Location
Chr. 1: 20183106 - 20196060 on Build GRCh38
UniGene ID
Hs.432503
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152376.4 NP_689589.1 2 266
XM_005245742.3 XP_005245799.1 2 770
XM_011540699.2 XP_011539001.1 2 474
GenBank mRNA AK058158.1 - 2 251
BC047428.1 - 2 261 5295853
BX648631.1 - 2 256
DB061550.1 - 2 266

View Full Product Details