Gene Transcripts

Gene Symbol
NUP35
Entrez Gene ID
129401
Gene Name
nucleoporin 35
Gene Aliases
MP-44, MP44, NP44, NUP53
Chromosome Location
Chr. 2: 183117490 - 183161684 on Build GRCh38
UniGene ID
Hs.180591
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287584.1 NP_001274513.1 7 760
NM_001287585.1 NP_001274514.1 7 780
NM_138285.4 NP_612142.2 6 657
NR_109856.1 5 515
XM_006712254.2 XP_006712317.1 6 622
XM_011510576.2 XP_011508878.1 8 888
XM_011510577.2 XP_011508879.1 8 1075
XM_017003308.1 XP_016858797.1 6 813
GenBank mRNA AF411516.1 - 6 657
AF514993.1 - 6 614
AK290760.1 - 6 569
AK298199.1 - 7 690
AK302350.1 - 7 729
BC035597.1 - 6 581
BC047029.1 - 6 644 5541023
BC061698.1 - 6 611 4132440
BI818393.1 - 6 617
BI821231.1 - 5 451

View Full Product Details