Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001257281.1 | NP_001244210.1 | 8 | 1023 | |
NM_152383.4 | NP_689596.4 | 8 | 1023 | ||
NR_046476.1 | 8 | 1023 | |||
GenBank mRNA | AK094293.1 | - | 8 | 1024 | |
AK295363.1 | - | 8 | 893 | ||
AL834174.1 | - | 7 | 621 | ||
BC026166.1 | - | 8 | 927 | 4822064 | |
BC036113.1 | - | 8 | 922 | 5297794 |