Gene Transcripts

Gene Symbol
LYPD6B
Entrez Gene ID
130576
Gene Name
LY6/PLAUR domain containing 6B
Gene Aliases
CT116, LYPD7
Chromosome Location
Chr. 2: 149038467 - 149215262 on Build GRCh38
UniGene ID
Hs.357567
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317002.1 NP_001303931.1 6 507
NM_001317003.1 NP_001303932.1 4 617
NM_001317004.1 NP_001303933.1 5 418
NM_001317005.1 NP_001303934.1 5 678
NM_001317006.1 NP_001303935.1 3 546
NM_177964.4 NP_808879.2 5 689
XM_005246306.4 XP_005246363.1 7 659
XM_006712279.2 XP_006712342.1 5 511
XM_006712280.3 XP_006712343.1 7 657
XM_006712281.3 XP_006712344.1 5 431
XM_011510620.2 XP_011508922.1 7 532
XM_011510623.2 XP_011508925.1 6 505
XM_011510625.1 XP_011508927.1 8 736
XM_017003364.1 XP_016858853.1 6 517
XM_017003365.1 XP_016858854.1 5 444
XM_017003366.1 XP_016858855.1 5 488
GenBank mRNA AF435957.1 - 5 418
BC018203.1 - 4 366 3861272
BC040176.2 - 5 689 4794338
BQ056975.1 - 3 283
CN359800.1 - 5 378

View Full Product Details