Gene Transcripts

Gene Symbol
CMBL
Entrez Gene ID
134147
Gene Name
carboxymethylenebutenolidase homolog
Gene Aliases
JS-1
Chromosome Location
Chr. 5: 10277595 - 10308056 on Build GRCh38
UniGene ID
Hs.192586
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138809.3 NP_620164.1 2 579
XM_017009034.1 XP_016864523.1 2 343
XM_017009035.1 XP_016864524.1 2 299
GenBank mRNA AJ278125.1 - 2 372
AK074197.1 - 2 370
AK315390.1 - 1 133
BC001573.1 - 2 285 3454199
BX647141.1 - 1 76

View Full Product Details