Gene Transcripts

Gene Symbol
ASB5
Entrez Gene ID
140458
Gene Name
ankyrin repeat and SOCS box containing 5
Gene Aliases
ASB-5
Chromosome Location
Chr. 4: 176213673 - 176277571 on Build GRCh38
UniGene ID
Hs.591712
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080874.3 NP_543150.1 5 766
XM_005262759.1 XP_005262816.1 7 1195
XM_011531616.2 XP_011529918.1 5 874
XM_011531617.2 XP_011529919.1 5 1457
GenBank mRNA AK096441.1 - 5 668
AK098693.1 - 5 671
AY057053.1 - 5 652
BC065710.1 - 5 673 4272258
BX647857.1 - 5 767

View Full Product Details