Gene Transcripts

Gene Symbol
TMEM37
Entrez Gene ID
140738
Gene Name
transmembrane protein 37
Gene Aliases
PR, PR1
Chromosome Location
Chr. 2: 119429912 - 119438520 on Build GRCh38
UniGene ID
Hs.26216
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183240.2 NP_899063.2 2 114
XM_005263598.4 XP_005263655.1 2 414
XM_006712300.3 XP_006712363.1 2 238
XM_011510659.2 XP_011508961.1 2 131
GenBank mRNA AF361356.1 - 2 80
BC046362.1 - 1 239 5221396
DR003856.1 - 2 121
DR005637.1 - 2 103

View Full Product Details