Gene Transcripts

Gene Symbol
ADAMTS14
Entrez Gene ID
140766
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 14
Gene Aliases
Chromosome Location
Chr. 10: 70672518 - 70762441 on Build GRCh38
UniGene ID
Hs.352156
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080722.4 NP_542453.2 7 1479
NM_139155.3 NP_631894.2 7 1488
XM_011539300.2 XP_011537602.1 7 1472
XM_011539301.2 XP_011537603.1 4 485
XM_011539302.2 XP_011537604.1 4 362
XM_011539303.2 XP_011537605.1 7 1476
XM_011539305.2 XP_011537607.1 7 1476
XM_011539306.2 XP_011537608.1 7 1476
XM_011539307.2 XP_011537609.1 7 1476
XM_011539308.2 XP_011537610.1 7 1476
GenBank mRNA AF358666.1 - 7 1182
AF366351.1 - 6 1109
AJ345098.1 - 7 1182

View Full Product Details