Gene Transcripts

Gene Symbol
ADAMTS14
Entrez Gene ID
140766
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 14
Gene Aliases
Chromosome Location
Chr. 10: 70672518 - 70762441 on Build GRCh38
UniGene ID
Hs.352156
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080722.4 NP_542453.2 20, 21 3365
NM_139155.3 NP_631894.2 20, 21 3374
XM_011539300.2 XP_011537602.1 17, 18 2848
XM_011539301.2 XP_011537603.1 17, 18 2371
XM_011539302.2 XP_011537604.1 17, 18 2248
XM_011539309.1 XP_011537611.1 13, 14 1958
GenBank mRNA AF358666.1 - 20, 21 3068
AF366351.1 - 19, 20 2995

View Full Product Details