Gene Transcripts

Gene Symbol
CACUL1
Entrez Gene ID
143384
Gene Name
CDK2 associated cullin domain 1
Gene Aliases
C10orf46, CAC1
Chromosome Location
Chr. 10: 118680982 - 118755246 on Build GRCh38
UniGene ID
Hs.420024
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153810.4 NP_722517.3 3, 4 1091
GenBank mRNA AJ544771.1 - 3, 4 875
AK097728.1 - 4, 5 811
AY743663.1 - 3, 4 1091
BC038294.2 - 3, 4 1091 4837841

View Full Product Details