Gene Transcripts

Gene Symbol
KDELC2
Entrez Gene ID
143888
Gene Name
KDEL motif containing 2
Gene Aliases
Chromosome Location
Chr. 11: 108472106 - 108498432 on Build GRCh38
UniGene ID
Hs.83286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153705.4 NP_714916.3 4 938
XM_011542621.2 XP_011540923.1 4 911
GenBank mRNA AF533708.1 - 4 897
AK125519.1 - 3 720
AK313534.1 - 4 800
AY358616.1 - 3 1007
BC036526.2 - 4 892 5266665

View Full Product Details