Gene Transcripts
- Gene Symbol
- LRRC74A
- Entrez Gene ID
- 145497
- Gene Name
- leucine rich repeat containing 74A
- Gene Aliases
- C14orf166B, LRRC74
- Chromosome Location
- Chr. 14: 76826370 - 76870302 on Build GRCh38
- UniGene ID
- Hs.147276
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_194287.2 |
NP_919263.2
|
4, 5 |
613 |
|
XM_011536479.1 |
XP_011534781.1
|
4, 5 |
981 |
|
XM_011536483.1 |
XP_011534785.1
|
4, 5 |
981 |
|
XM_011536487.1 |
XP_011534789.1
|
4, 5 |
981 |
|
XM_017021032.1 |
XP_016876521.1
|
4, 5 |
879 |
|
XM_017021033.1 |
XP_016876522.1
|
4, 5 |
981 |
|
XM_017021034.1 |
XP_016876523.1
|
4, 5 |
625 |
|
XM_017021035.1 |
XP_016876524.1
|
4, 5 |
776 |
|
XM_017021036.1 |
XP_016876525.1
|
4, 5 |
981 |
|
XM_017021037.1 |
XP_016876526.1
|
4, 5 |
1683 |
|
XM_017021038.1 |
XP_016876527.1
|
4, 5 |
517 |
|
XM_017021039.1 |
XP_016876528.1
|
3, 4 |
439 |
|
XM_017021040.1 |
XP_016876529.1
|
4, 5 |
438 |
|
GenBank mRNA |
AK125722.1 |
- |
5, 6 |
836 |
|
AK301762.1 |
- |
5, 6 |
759 |
|
BC062799.1 |
- |
5, 6 |
1173 |
|
BC121159.2 |
- |
4, 5 |
461 |
40121413 |
BC121160.2 |
- |
4, 5 |
461 |
40121414 |
View Full Product Details