Gene Transcripts

Gene Symbol
NFXL1
Entrez Gene ID
152518
Gene Name
nuclear transcription factor, X-box binding like 1
Gene Aliases
CDZFP, HOZFP, OZFP, URCC5
Chromosome Location
Chr. 4: 47847233 - 47914667 on Build GRCh38
UniGene ID
Hs.646325
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278623.1 NP_001265552.1 14 1738
NM_001278624.1 NP_001265553.1 14 1876
NM_152995.5 NP_694540.3 14 1778
NR_103795.1 14 1738
GenBank mRNA AB085695.1 - 14 1734
AB181916.1 - 14 1858
AK131285.1 - 12 1324
AK302440.1 - 14 1736
BC051193.1 - 14 1709 5271401

View Full Product Details