Gene Transcripts

Gene Symbol
FREM1
Entrez Gene ID
158326
Gene Name
FRAS1 related extracellular matrix 1
Gene Aliases
BNAR, C9orf143, C9orf145, C9orf154, MOTA, TILRR, TRIGNO2
Chromosome Location
Chr. 9: 14734666 - 14911653 on Build GRCh38
UniGene ID
Hs.50850
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177704.1 NP_001171175.1 11 1800
NM_144966.5 NP_659403.4 35 6919
XM_005251382.3 XP_005251439.1 34 7167
XM_005251384.4 XP_005251441.1 13 2213
XM_006716729.3 XP_006716792.1 14 2458
XM_011517758.2 XP_011516060.1 11 1849
XM_017014316.1 XP_016869805.1 34 7194
XM_017014317.1 XP_016869806.1 35 6737
XM_017014318.1 XP_016869807.1 35 7492
XM_017014319.1 XP_016869808.1 36 7396
XM_017014320.1 XP_016869809.1 35 7323
XM_017014321.1 XP_016869810.1 34 6986
XM_017014322.1 XP_016869811.1 34 6780
XM_017014323.1 XP_016869812.1 34 6453
XM_017014324.1 XP_016869813.1 33 7044
XM_017014325.1 XP_016869814.1 34 7194
XM_017014326.1 XP_016869815.1 33 6010
XM_017014327.1 XP_016869816.1 30 5379
GenBank mRNA AB160987.1 - 33 6103
AK058190.1 - 8 954
BC031064.1 - 11 1800
BX537574.1 - 28 5494
BX641104.1 - 5 6424

View Full Product Details