Gene Transcripts

Gene Symbol
FAM71D
Entrez Gene ID
161142
Gene Name
family with sequence similarity 71 member D
Gene Aliases
C14orf54
Chromosome Location
Chr. 14: 67188365 - 67228550 on Build GRCh38
UniGene ID
Hs.412818
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173526.3 NP_775797.2 6 1386
XM_011536500.1 XP_011534802.1 6 1350
XM_011536502.1 XP_011534804.1 6 1285
XM_011536503.2 XP_011534805.1 5 2327
GenBank mRNA AK093449.1 - 6 1350
BC050401.1 - 7 1252 5745029

View Full Product Details