Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304787.1 | NP_001291716.1 | 3 | 569 | |
NM_153360.2 | NP_699191.1 | 2 | 398 | ||
NR_130908.1 | 2 | 398 | |||
GenBank mRNA | AK074647.1 | - | 2 | 353 | |
AK293299.1 | - | 3 | 528 | ||
BC101758.1 | - | 2 | 250 | 8069264 | |
BC101760.1 | - | 2 | 250 | 8069266 | |
DA665037.1 | - | 2 | 398 |