Gene Transcripts

Gene Symbol
SMIM14
Entrez Gene ID
201895
Gene Name
small integral membrane protein 14
Gene Aliases
C4orf34
Chromosome Location
Chr. 4: 39547297 - 39638998 on Build GRCh38
UniGene ID
Hs.205952
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317896.1 NP_001304825.1 3, 4 509
NM_001317897.1 NP_001304826.1 2, 3 218
NM_174921.2 NP_777581.1 2, 3 364
XM_011513640.2 XP_011511942.1 2, 3 497
XM_017007862.1 XP_016863351.1 3, 4 413
XM_017007863.1 XP_016863352.1 2, 3 254
GenBank mRNA AK023351.1 - 2, 3 227
BC008502.1 - 2, 3 227 4295121
BQ049060.1 - 2, 3 209
HY004916.1 - 3, 4 410
HY155982.1 - 2, 3 365

View Full Product Details