Gene Transcripts

Gene Symbol
SLC17A8
Entrez Gene ID
246213
Gene Name
solute carrier family 17 member 8
Gene Aliases
DFNA25, VGLUT3
Chromosome Location
Chr. 12: 100357079 - 100422059 on Build GRCh38
UniGene ID
Hs.116871
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145288.1 NP_001138760.1 2, 3 665
NM_139319.2 NP_647480.1 2, 3 665
GenBank mRNA AJ459241.1 - 2, 3 528
AK128319.1 - 2, 3 665
BC117229.1 - 2, 3 376 40125780
BC143396.1 - 2, 3 376 9051903

View Full Product Details