Gene Transcripts

Gene Symbol
ETFBKMT
Entrez Gene ID
254013
Gene Name
electron transfer flavoprotein beta subunit lysine methyltransferase
Gene Aliases
C12orf72, ETFB-KMT, METTL20
Chromosome Location
Chr. 12: 31647160 - 31669082 on Build GRCh38
UniGene ID
Hs.740628
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135863.1 NP_001129335.1 4 861
NM_001135864.1 NP_001129336.1 4 842
NM_173802.3 NP_776163.1 4 922
GenBank mRNA AK290248.1 - 4 843
AL832339.1 - 4 848
BC039107.1 - 5 1313 4837913
BC039535.1 - 4 922 5744381

View Full Product Details