Gene Transcripts

Gene Symbol
LINC00521
Entrez Gene ID
256369
Gene Name
long intergenic non-protein coding RNA 521
Gene Aliases
C14orf48, c14_5713
Chromosome Location
Chr. 14: 93997270 - 94011695 on Build GRCh38
UniGene ID
Hs.143845
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024182.1 6 684
NR_024183.1 4 712
NR_024184.1 4 712
GenBank mRNA AK097741.1 - 4 707
BC021728.2 - 4 711
BC031252.1 - 6 684

View Full Product Details