Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_198471.2 | NP_940873.2 | 9 | 2236 | |
XM_006722718.3 | XP_006722781.1 | 9 | 2355 | ||
XM_011527884.2 | XP_011526186.1 | 9 | 2612 | ||
XM_017026563.1 | XP_016882052.1 | 8 | 2372 | ||
GenBank mRNA | AB284126.1 | - | 9 | 2236 | |
AK128815.1 | - | 9 | 2236 | ||
BC066124.1 | - | 9 | 2236 | 30345663 | |
BC066775.1 | - | 9 | 2236 | 30333203 |