Gene Transcripts

Gene Symbol
TMED8
Entrez Gene ID
283578
Gene Name
transmembrane p24 trafficking protein family member 8
Gene Aliases
FAM15B
Chromosome Location
Chr. 14: 77335021 - 77377384 on Build GRCh38
UniGene ID
Hs.200413
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_213601.1 NP_998766.1 5 682
XM_017021223.1 XP_016876712.1 5 1028
XM_017021224.1 XP_016876713.1 4 581
XM_017021225.1 XP_016876714.1 5 3143
GenBank mRNA AK095650.1 - 5 726
AY598329.1 - 5 682
BC101510.1 - 5 738 8069016
BX248262.1 - 1 107

View Full Product Details