Gene Transcripts

Gene Symbol
ZFP69
Entrez Gene ID
339559
Gene Name
ZFP69 zinc finger protein
Gene Aliases
ZFP69A, ZKSCAN23A, ZNF642, ZSCAN54A
Chromosome Location
Chr. 1: 40477220 - 40496343 on Build GRCh38
UniGene ID
Hs.434385
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320178.1 NP_001307107.1 6 802
NM_001320179.1 NP_001307108.1 6 1096
NM_198494.2 NP_940896.2 6 799
XM_005270809.4 XP_005270866.1 6 1529
XM_006710605.3 XP_006710668.1 6 1509
XM_006710606.3 XP_006710669.1 6 1526
GenBank mRNA AK122618.1 - 6 799
BC041873.1 - 6 1512 5271835
BC144174.1 - 5 548
CV374692.1 - 4 375
CX871163.1 - 3 185

View Full Product Details