Gene Transcripts

Gene Symbol
FAM221A
Entrez Gene ID
340277
Gene Name
family with sequence similarity 221 member A
Gene Aliases
C7orf46
Chromosome Location
Chr. 7: 23680114 - 23703249 on Build GRCh38
UniGene ID
Hs.592178
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127364.2 NP_001120836.1 4 557
NM_001127365.2 NP_001120837.1 3 383
NM_001300932.1 NP_001287861.1 3 383
NM_199136.4 NP_954587.2 4 557
XM_011515369.2 XP_011513671.1 4 552
XM_011515370.2 XP_011513672.1 4 552
XM_011515371.2 XP_011513673.1 4 552
XM_011515372.2 XP_011513674.1 4 697
XM_017012134.1 XP_016867623.1 4 552
XM_017012135.1 XP_016867624.1 4 697
GenBank mRNA AK127774.1 - 5 3206
AL832127.1 - 4 493
BC026000.1 - 3 315
BC042034.1 - 4 538
BC063130.1 - 4 471 30382529
BI549491.1 - 4 542
BU958830.1 - 3 315

View Full Product Details