Gene Transcripts

Gene Symbol
ZCCHC16
Entrez Gene ID
340595
Gene Name
zinc finger CCHC-type containing 16
Gene Aliases
Mar4, Mart4
Chromosome Location
Chr. X: 112083025 - 112457245 on Build GRCh38
UniGene ID
Hs.368140
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004308.2 NP_001004308.2 3 766
GenBank mRNA AK128465.1 - 3 766
BC137430.1 - 1 559 9021428
BC137433.1 - 1 559 9021431

View Full Product Details