Gene Transcripts

Gene Symbol
KRT27
Entrez Gene ID
342574
Gene Name
keratin 27
Gene Aliases
K25IRS3, KRT25C
Chromosome Location
Chr. 17: 40776808 - 40782534 on Build GRCh38
UniGene ID
Hs.59363
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181537.3 NP_853515.2 3 613
GenBank mRNA AJ564206.1 - 3 613

View Full Product Details