Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145664.1 | NP_001139136.1 | 11 | 1099 | |
XM_011511771.2 | XP_011510073.1 | 12 | 1637 | ||
XM_011511772.1 | XP_011510074.1 | 14 | 1311 | ||
XM_011511773.1 | XP_011510075.1 | 10 | 982 | ||
XM_011511776.1 | XP_011510078.1 | 13 | 1270 | ||
XM_011511777.1 | XP_011510079.1 | 8 | 1640 | ||
XM_011511778.1 | XP_011510080.1 | 8 | 977 | ||
XM_017004851.1 | XP_016860340.1 | 14 | 1321 | ||
XM_017004852.1 | XP_016860341.1 | 11 | 1108 | ||
GenBank mRNA | AK124976.1 | - | 7 | 2154 | |
AK131289.1 | - | 13 | 1277 | ||
AK298610.1 | - | 11 | 1099 |