Gene Transcripts

Gene Symbol
OR52H1
Entrez Gene ID
390067
Gene Name
olfactory receptor family 52 subfamily H member 1
Gene Aliases
OR11-41, OR11-45
Chromosome Location
Chr. 11: 5544561 - 5545523 on Build GRCh38
UniGene ID
Hs.554540
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005289.1 NP_001005289.1 1 492
GenBank mRNA BC136998.1 - 1 518

View Full Product Details