Gene Transcripts

Gene Symbol
OR4N5
Entrez Gene ID
390437
Gene Name
olfactory receptor family 4 subfamily N member 5
Gene Aliases
Chromosome Location
Chr. 14: 20143736 - 20144662 on Build GRCh38
UniGene ID
Hs.553766
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004724.1 NP_001004724.1 1 858

View Full Product Details