Gene Transcripts

Gene Symbol
OR2A25
Entrez Gene ID
392138
Gene Name
olfactory receptor family 2 subfamily A member 25
Gene Aliases
OR2A24P, OR2A25P, OR2A27
Chromosome Location
Chr. 7: 144074220 - 144075152 on Build GRCh38
UniGene ID
Hs.553787
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004488.1 NP_001004488.1 1 569
GenBank mRNA BC136814.1 - 1 607
BC136829.1 - 1 607 9020820

View Full Product Details