Gene Transcripts

Gene Symbol
STK32C
Entrez Gene ID
282974
Gene Name
serine/threonine kinase 32C
Gene Aliases
PKE, YANK3
Chromosome Location
Chr. 10: 132207482 - 132332559 on Build GRCh38
UniGene ID
Hs.469002
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318878.1 NP_001305807.1 10, 11 1429
NM_001318879.1 NP_001305808.1 10, 11 1290
NM_173575.3 NP_775846.2 10, 11 1391
XM_011539688.1 XP_011537990.1 10, 11 1462
XM_011539690.2 XP_011537992.1 10, 11 1377
XM_011539693.2 XP_011537995.1 10, 11 1395
XM_011539694.1 XP_011537996.1 10, 11 1129
XM_011539695.1 XP_011537997.1 10, 11 1330
XM_011539696.1 XP_011537998.1 10, 11 1137
GenBank mRNA AK098366.1 - 8, 9 1418
AK302102.1 - 10, 11 1376
AY098866.1 - 10, 11 1391
BC045760.1 - 10, 11 1280 4831007

View Full Product Details