Gene Transcripts

Gene Symbol
PDXP
Entrez Gene ID
57026
Gene Name
pyridoxal phosphatase
Gene Aliases
CIN, PLP, dJ37E16.5
Chromosome Location
Chr. 22: 37658730 - 37666932 on Build GRCh38
UniGene ID
Hs.632762
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020315.4 NP_064711.1 2 713
GenBank mRNA AK057265.1 - 2 279
AY125047.1 - 2 660
BC000320.2 - 2 712 2821743
BC009756.2 - 2 302
BC018844.2 - 2 681
BC064922.1 - 2 713 6141538

View Full Product Details