Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005804.3 | NP_005795.2 | 2 | 225 | |
NR_046366.2 | 2 | 225 | |||
XM_006722606.3 | XP_006722669.1 | 2 | 165 | ||
XM_011527620.1 | XP_011525922.1 | 2 | 244 | ||
XM_011527621.2 | XP_011525923.1 | 1 | 111 | ||
GenBank mRNA | AB254849.1 | - | 1 | 107 | |
AK293911.1 | - | 2 | 183 | ||
AK301847.1 | - | 3 | 308 | ||
BC001009.2 | - | 2 | 162 | 2820926 | |
BC032128.2 | - | 2 | 171 | 5017293 | |
DA432925.1 | - | 2 | 226 | ||
DB032777.1 | - | 3 | 318 | ||
U90426.1 | - | 2 | 202 |