Gene Transcripts

Gene Symbol
AMELX
Entrez Gene ID
265
Gene Name
amelogenin, X-linked
Gene Aliases
AI1E, AIH1, ALGN, AMG, AMGL, AMGX
Chromosome Location
Chr. X: 11293413 - 11300761 on Build GRCh38
UniGene ID
Hs.654436
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142.2 NP_001133.1 4 190
NM_182680.1 NP_872621.1 5 232
NM_182681.1 NP_872622.1 3 142
XM_017029404.1 XP_016884893.1 3 1291
GenBank mRNA AF436849.1 - 5 232
BC069118.1 - 4 190 7216972
BC074951.2 - 4 169 30915277
M86932.1 - 4 190

View Full Product Details