Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001229.4 | NP_001220.2 | 2 | 521 | |
NM_001278054.1 | NP_001264983.1 | 2 | 521 | ||
NM_032996.3 | NP_127463.2 | 2 | 352 | ||
NR_102732.1 | 2 | 521 | |||
XM_005246014.2 | XP_005246071.1 | 2 | 205 | ||
XM_011542273.2 | XP_011540575.1 | 2 | 315 | ||
GenBank mRNA | AB015653.1 | - | 2 | 297 | |
AB020979.1 | - | 2 | 276 | ||
AB209147.1 | - | 2 | 521 | ||
AF093130.1 | - | 2 | 276 | ||
AF110376.1 | - | 2 | 276 | ||
AK292080.1 | - | 2 | 293 | ||
AK292111.1 | - | 2 | 297 | ||
AK303743.1 | - | 2 | 288 | ||
AY732490.2 | - | 2 | 276 | ||
BC002452.2 | - | 2 | 282 | 3343958 | |
BC006463.1 | - | 2 | 371 | 3542075 | |
BT006911.1 | - | 2 | 276 | ||
DA078665.1 | - | 2 | 253 | ||
HY027003.1 | - | 2 | 315 | ||
U60521.1 | - | 2 | 321 |