Gene Transcripts

Gene Symbol
NDUFB6
Entrez Gene ID
4712
Gene Name
NADH:ubiquinone oxidoreductase subunit B6
Gene Aliases
B17, CI
Chromosome Location
Chr. 9: 32553526 - 32573184 on Build GRCh38
UniGene ID
Hs.493668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199987.1 NP_001186916.1 1 181
NM_002493.4 NP_002484.1 1 181
NM_182739.2 NP_877416.1 1 181
GenBank mRNA AF035840.1 - 1 125
AF067167.1 - 1 154
AK289702.1 - 1 135
AW160372.1 - 1 112
BC009801.1 - 1 160 4334414
BC016708.1 - 1 195
BC029247.1 - 1 126 5171627
BI669356.1 - 1 160
BP208846.1 - 1 179
CB140469.1 - 1 181
CR456920.1 - 1 57

View Full Product Details